Invitrogen
Manufacturer Code:MA527353
Catalog # PIMA527353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 2-150 of POLR2H produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase II subunit H; DNA-directed RNA polymerases I, II, and III 17.1 kDa polypeptide; DNA-directed RNA polymerases I, II, and III subunit RPABC3; hRPB8; polymerase (RNA) II (DNA directed) polypeptide H; polymerase (RNA) II subunit H; RNA polymerases I, II, and III subunit ABC3; RPABC3 RPB17 RPB8 hsRPB8 DNA-directed RNA polymerase II subunit H DNA-directed RNA polymerases I II and III 17.1 kDa polypeptide DNA-directed RNA polymerases I II and III subunit RPABC3 RNA polymerases I II and III subunit ABC3 RPB8 homolog hRPB8; RPB17; RPB8 homolog
Gene Aliases: POLR2H; RPABC3; RPB17; RPB8
UniProt ID: (Human) P52434
Entrez Gene ID: (Human) 5437
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.