Proteintech
Manufacturer Code:150861AP150UL
Catalog # 50-172-9966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | POLR2H Fusion Protein Ag7166 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase II subunit H; DNA-directed RNA polymerases I, II, and III 17.1 kDa polypeptide; DNA-directed RNA polymerases I, II, and III subunit RPABC3; hRPB8; hRPB8 hsRPB8 POLR2H RPABC3 RPB17 RPB8 RPB8 homolog; polymerase (RNA) II (DNA directed) polypeptide H; polymerase (RNA) II subunit H; RNA polymerases I, II, and III subunit ABC3; RPB17; RPB8 homolog
Gene Aliases: POLR2H; RGD1561203; RPABC3; RPB17; RPB8
UniProt ID: (Human) P52434, (Mouse) Q923G2
Entrez Gene ID: (Human) 5437, (Mouse) 245841, (Rat) 498109
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.