OriGene
Manufacturer Code:CF807758
Catalog # 50-166-8463
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F2 |
Immunogen | Full length human recombit protein of human POLR2F produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase II subunit F; DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide; DNA-directed RNA polymerases I, II, and III subunit RPABC2; HRBP14.4 POLRF RPABC14.4 RPABC2 RPB14.4 RPB6 RPC15; polymerase (RNA) II (DNA directed) polypeptide F; polymerase (RNA) II subunit F; RNA Polymerase II subunit 14.4 kD; RNA polymerases I, II, and III subunit ABC2; RPABC14.4; RPB14.4; RPB6 homolog; RPC15
Gene Aliases: HRBP14.4; POLR2F; POLRF; RPABC14.4; RPABC2; RPB14.4; RPB6; RPC15
UniProt ID: (Human) P61218
Entrez Gene ID: (Human) 5435
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.