Invitrogen
Manufacturer Code:PA589048
Catalog # PIPA589048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-127 of human POLR2F |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase II subunit F; DNA-directed RNA polymerases I, II, and III subunit RPABC2; RNA Pol II; RNA polymerases I, II, and III subunit ABC2; RPB6 homolog
Gene Aliases: 1810060D16Rik; Polr2f
UniProt ID: (Mouse) P61219
Entrez Gene ID: (Mouse) 69833
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.