Invitrogen
Manufacturer Code:PA5103803
Catalog # PIPA5103803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RPAB2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810060D16Rik DNA-directed RNA polymerase II subunit F DNA-directed RNA polymerases I II and III 14.4 kDa polypeptide DNA-directed RNA polymerases I II and III subunit RPABC2 HRBP14.4 hsRPB6 Polr2f POLRF polymerase (RNA) II (DNA directed) polypeptide F polymerase (RNA) II subunit F polymerase II RNA Pol II RNA Polymerase II subunit 14.4 kD RNA polymerase II subunit F RNA polymerases I II and III subunit ABC2 RPABC14.4 RPABC2 RPB14.4 RPB6 RPB6 homolog RPC15; DNA-directed RNA polymerase II subunit F; DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide; DNA-directed RNA polymerases I, II, and III subunit RPABC2; polymerase (RNA) II (DNA directed) polypeptide F; polymerase (RNA) II subunit F; polymerase II; RNA Pol II; RNA Polymerase II subunit 14.4 kD; RNA polymerases I, II, and III subunit ABC2; RPABC14.4; RPB14.4; RPB6 homolog; RPC15
Gene Aliases: 1810060D16Rik; HRBP14.4; POLR2F; POLRF; RPABC14.4; RPABC2; RPB14.4; RPB6; RPC15
UniProt ID: (Human) P61218, (Mouse) P61219, (Rat) O88828
Entrez Gene ID: (Human) 5435, (Mouse) 69833, (Rat) 83503
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.