Novus Biologicals
Manufacturer Code:NBP29358020UL
Catalog # NB029030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human POLR2F (NP_068809.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase II subunit F; DNA-directed RNA polymerase II subunit F DNA-directed RNA polymerases I II and III 14.4 kDa polypeptide HRBP14.4 II and III subunit RPABC2 polymerase (RNA) II (DNA directed) polypeptide F RNA Polymerase II subunit 14.4 kD RNA polymerases I II and III subunit ABC2 RPB6 RPB6 homolog; DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide; DNA-directed RNA polymerases I, II, and III subunit RPABC2; polymerase (RNA) II (DNA directed) polypeptide F; polymerase (RNA) II subunit F; RNA Polymerase II subunit 14.4 kD; RNA polymerases I, II, and III subunit ABC2; RPABC14.4; RPB14.4; RPB6 homolog; RPC15
Gene Aliases: HRBP14.4; POLR2F; POLRF; RPABC14.4; RPABC2; RPB14.4; RPB6; RPC15
UniProt ID: (Human) P61218
Entrez Gene ID: (Human) 5435
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.