Novus Biologicals
Manufacturer Code:NBP273518B
Catalog # NBP273518B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B5 |
Immunogen | Full length recombinant protein of human POLR2E (NP_002686) produced in HEK293T cells. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DNA directed RNA polymerase II 23 kda polypeptide; DNA directed RNA polymerase II 23 kda polypeptide DNA-directed RNA polymerase II 23 kDa polypeptide DNA-directed RNA polymerase II subunit E DNA-directed RNA polymerases I II and III subunit RPABC1 hRPB25 hsRPB5 polymerase (RNA) II (DNA directed) polypeptide E (25kD) polymerase (RNA) II (DNA directed) polypeptide E 25kDa RPABC1 RPB5 RPB5 homolog XAP4RNA polymerases I II and III subunit ABC1; DNA-directed RNA polymerase II 23 kDa polypeptide; DNA-directed RNA polymerase II subunit E; DNA-directed RNA polymerase subunit RPABC1; DNA-directed RNA polymerases I, II, and III subunit RPABC1; polymerase (RNA) II (DNA directed) polypeptide E, 25kDa; polymerase (RNA) II subunit E; RNA polymerases I, II, and III subunit ABC1; RPB5 homolog; XAP4
Gene Aliases: hRPB25; hsRPB5; POLR2E; RPABC1; RPB5; XAP4
UniProt ID: (Human) P19388
Entrez Gene ID: (Human) 5434
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.