Invitrogen
Manufacturer Code:MA535851
Catalog # PIMA535851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1541 |
Immunogen | A phospho specific peptide corresponding to residues surrounding S5 of human POLR2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 220kDa BOS_18773 DNA-directed RNA polymerase II subunit A DNA-directed RNA polymerase II subunit RPB1 DNA-directed RNA polymerase III largest subunit EC 2.7.7.6 hRPB220 hsRPB1 MGC75453 Pol II S2p POLR2 POLR2A POLRA POLYMERASE II RNA RNA POL2 RNA polymerase II subunit B1 RNA-directed RNA polymerase II subunit RPB1 RP02 Rpb1 RPB220 RPBh1 Rpii215 RpIILS RPO2 RPO21 Rpo2-1 RPOL2 SUA8; DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit; DNA-directed RNA polymerase II subunit A; DNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; polymerase (RNA) II (DNA directed) polypeptide A, 220kDa; polymerase (RNA) II subunit A; RNA polymerase II 1; RNA polymerase II subunit B1; RNA-directed RNA polymerase II subunit RPB1
Gene Aliases: 220kDa; hRPB220; hsRPB1; POLR2; POLR2A; POLRA; RPB1; RPBh1; Rpii215; RpIILS; RPO2; Rpo2-1; RPOL2
UniProt ID: (Human) P24928, (Mouse) P08775
Entrez Gene ID: (Human) 5430, (Mouse) 20020
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.