Abnova
Manufacturer Code:MAB19497
Catalog # 89-336-022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | AH-16 |
Immunogen | A synthetic peptide corresponding to human POLR2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit; DNA-directed RNA polymerase II subunit A; DNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; MGC75453 POLR2 POLRA RPB1 RPBh1 RPO2 RPOL2 RpIILS hRPB220 hsRPB1; polymerase (RNA) II (DNA directed) polypeptide A, 220kDa; polymerase (RNA) II subunit A; RNA polymerase II subunit B1; RNA-directed RNA polymerase II subunit RPB1
Gene Aliases: hRPB220; hsRPB1; POLR2; POLR2A; POLRA; RPB1; RPBh1; RpIILS; RPO2; RPOL2
UniProt ID: (Human) P24928
Entrez Gene ID: (Human) 5430
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.