Proteintech
Manufacturer Code:206551AP150UL
Catalog # 50-173-3016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kd subunit; DNA-directed RNA polymerase II subunit A; DNA-directed RNA polymerase II subunit RPB1; DNA-directed RNA polymerase III largest subunit; hRPB220 hsRPB1 Pol II POLR2 POLR2A POLRA RNA polymerase II subunit B1 RPB1 RPBh1 RpIILS RPO2 RPOL2; polymerase (RNA) II (DNA directed) polypeptide A, 220kDa; polymerase (RNA) II subunit A; RNA polymerase II subunit B1; RNA-directed RNA polymerase II subunit RPB1
Gene Aliases: hRPB220; hsRPB1; POLR2; POLR2A; POLRA; RPB1; RPBh1; RpIILS; RPO2; RPOL2
UniProt ID: (Human) P24928
Entrez Gene ID: (Human) 5430
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.