Proteintech
Manufacturer Code:159231AP150UL
Catalog # 50-173-0576
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | POLR1C Fusion Protein Ag8746 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AC40; AC40 POLR1C POLR1E RPA39 RPA40 RPA5 RPAC1 RPC40; DNA-directed RNA polymerase I subunit C; DNA-directed RNA polymerases I and III 40 kDa polypeptide; DNA-directed RNA polymerases I and III subunit RPAC1; polymerase (RNA) I polypeptide C; polymerase (RNA) I polypeptide C, 30kDa; polymerase (RNA) I subunit C; RNA polymerase 1-1; RNA polymerase 1-1 (40 kDa subunit); RNA polymerase I subunit; RNA polymerases I and III subunit AC1; RPA39; RPA40; RPC40
Gene Aliases: 40kDa; AA409007; AA959927; Ac2-127; AC40; AL024089; HLD11; POLR1C; POLR1E; RPA39; RPA40; RPA5; RPAC1; RPC40; Rpo1-1; TCS3
UniProt ID: (Human) O15160, (Mouse) P52432
Entrez Gene ID: (Human) 9533, (Rat) 301246, (Mouse) 20016
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.