Invitrogen
Manufacturer Code:PA539139
Catalog # PIPA539139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human POLR1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A127; DNA-directed RNA polymerase I 135 kDa polypeptide; DNA-directed RNA polymerase I 135kDa polypeptide; DNA-directed RNA polymerase I subunit RPA2; polymerase (RNA) I polypeptide B; polymerase (RNA) I polypeptide B, 128kDa; polymerase (RNA) I subunit B; RNA polymerase 1-2; RNA polymerase 1-2, 128 kDa subunit; RNA polymerase I (127 kDa subunit); RNA polymerase I polypeptide B; RNA polymerase I subunit 2; RPA135
Gene Aliases: 128kDa; D630020H17Rik; POLR1B; RPA116; RPA135; RPA2; Rpo1-2
UniProt ID: (Human) Q9H9Y6, (Mouse) P70700, (Rat) O54888
Entrez Gene ID: (Human) 84172, (Mouse) 20017, (Rat) 83582
Molecular Function: DNA-directed RNA polymerase RNA binding protein nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.