Invitrogen
Manufacturer Code:PA5115130
Catalog # PIPA5115130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DNA Polymerase theta corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromosome aberrations occurring spontaneously protein 1; DNA polymerase eta; DNA polymerase eta DNA polymerase theta EC 2.7.7.7 POLH POLQ polymerase (DNA directed) theta polymerase (DNA) theta PRO0327; DNA polymerase theta; polymerase (DNA directed), theta; polymerase (DNA) theta
Gene Aliases: A430110D14Rik; Chaos1; POLH; POLQ; PRO0327
UniProt ID: (Human) O75417, (Mouse) Q8CGS6
Entrez Gene ID: (Human) 10721, (Rat) 288079, (Mouse) 77782
Molecular Function:
DNA binding protein
DNA-directed DNA polymerase
exodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.