Abnova
Manufacturer Code:PAB15622
Catalog # 89-117-692
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at internal region of human POLQ. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised aganist synthetic peptide of POLQ.
For Research Use Only
Protein Aliases: DKFZp781A0112; DNA polymerase eta; DNA polymerase theta; POLH; polymerase (DNA directed), theta; polymerase (DNA) theta; PRO0327
Gene Aliases: POLH; POLQ; PRO0327
UniProt ID: (Human) O75417
Entrez Gene ID: (Human) 10721
Molecular Function: DNA binding protein DNA-directed DNA polymerase exodeoxyribonuclease hydrolase nuclease nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.