Novus Biologicals
Manufacturer Code:NBP299015FR
Catalog # NB057287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human POLG2. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA polymerase gamma accessory 55 kDa subunit; DNA polymerase gamma accessory 55 kDa subunit DNA polymerase subunit gamma-2 mitochondrial EC 2.7.7.7 HP55 Mitochondrial DNA polymerase accessory subunit mitochondrial DNA polymerase subunit gamma-2 mitochondrial DNA polymerase accessory subunit MtPolB MTPOLBPOLG-BETA p55 PEOA4 POLB POLGB PolG-beta polymerase (DNA directed) gamma 2 accessory subunit; DNA polymerase subunit gamma-2, mitochondrial; Mitochondrial DNA polymerase accessory subunit; mitochondrial DNA polymerase subunit gamma-2; mitochondrial DNA polymerase, accessory subunit; MtPolB; p55; PolG-beta; polymerase (DNA directed), gamma 2, accessory subunit; polymerase (DNA) gamma 2, accessory subunit
Gene Aliases: HP55; MTPOLB; PEOA4; POLB; POLG-BETA; POLG2; POLGB
UniProt ID: (Human) Q9UHN1
Entrez Gene ID: (Human) 11232
Molecular Function: aminoacyl-tRNA synthetase ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.