Invitrogen
Manufacturer Code:MA535359
Catalog # PIMA535359
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0752 |
Immunogen | A synthesized peptide derived from human DNA Polymerase beta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: A430088C08Rik BOS_23903 DNA polymerase beta MGC125976 POL B Polb polymerase B wu:fa97a05 XELAEV_18020165mg zgc:109924; DNA pol beta; DNA polymerase beta; DNA polymerase beta subunit; DNA-directed DNA polymerase beta; polymerase (DNA directed), beta; polymerase (DNA) beta; polymerase-beta
Gene Aliases: POLB
UniProt ID: (Human) P06746, (Rat) P06766
Entrez Gene ID: (Human) 5423, (Rat) 29240
Molecular Function:
DNA binding protein
DNA-directed DNA polymerase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.