Invitrogen
Manufacturer Code:PA534684
Catalog # PIPA534684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 330 of DNA Polymerase beta (Uniprot IDNo.P06746) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA pol beta; DNA pol beta DNA polymerase beta DNA polymerase beta subunit mutant DNA polymerase beta POLB POL B polymerase B; DNA polymerase beta; DNA polymerase beta subunit; polymerase (DNA directed), beta; polymerase (DNA) beta
Gene Aliases: POLB
UniProt ID: (Human) P06746
Entrez Gene ID: (Human) 5423
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.