Abnova
Manufacturer Code:PAB27052
Catalog # 89-128-702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human POLB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of POLB.
For Research Use Only
Protein Aliases: DNA pol beta; DNA polymerase beta; DNA polymerase beta subunit; MGC125976; polymerase (DNA directed), beta; polymerase (DNA) beta
Gene Aliases: POLB
UniProt ID: (Human) P06746
Entrez Gene ID: (Human) 5423
Molecular Function:
DNA binding protein
DNA-directed DNA polymerase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.