Invitrogen
Manufacturer Code:PA570796
Catalog # PIPA570796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of mouse POLA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA polymerase alpha 2, 68 kDa; DNA polymerase alpha 70 kDa subunit; DNA polymerase alpha subunit B; DNA polymerase alpha subunit II; DNA polymerase subunit II; DNA-directed DNA polymerase alpha 2; Ese1L; polymerase (DNA directed), alpha 2 (70kD subunit); polymerase (DNA directed), alpha 2, accessory subunit; polymerase (DNA) alpha 2, accessory subunit; polymerase (DNA-directed), alpha (70kD)
Gene Aliases: AI573378; POLA2
UniProt ID: (Human) Q14181, (Bovine) Q58D13, (Mouse) P33611
Entrez Gene ID: (Dog) 476025, (Human) 23649, (Bovine) 514793, (Rabbit) 100355173, (Horse) 100051254, (Mouse) 18969, (Guinea pig) 100727306, (Rat) 85242
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.