Invitrogen
Manufacturer Code:PA5103789
Catalog # PIPA5103789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human POLA2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI573378 DNA polymerase alpha 2 68 kDa DNA polymerase alpha 2 accessory subunit DNA polymerase alpha 70 kDa subunit DNA polymerase alpha subunit B DNA polymerase alpha subunit II DNA polymerase subunit II DNA-directed DNA polymerase alpha 2 Ese1L Pola2 polymerase (DNA directed) alpha 2 polymerase (DNA directed) alpha 2 (70kD subunit) polymerase (DNA directed) alpha 2 accessory subunit polymerase (DNA) alpha 2 accessory subunit polymerase (DNA-directed) alpha (70kD); DNA polymerase alpha 70 kDa subunit; DNA polymerase alpha subunit B; polymerase (DNA directed), alpha 2 (70kD subunit); polymerase (DNA directed), alpha 2, accessory subunit; polymerase (DNA) alpha 2, accessory subunit; polymerase (DNA-directed), alpha (70kD)
Gene Aliases: POLA2
UniProt ID: (Human) Q14181
Entrez Gene ID: (Human) 23649
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.