Invitrogen
Manufacturer Code:PA551784
Catalog # PIPA551784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human POLA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA polymerase alpha 1 catalytic subunit; DNA polymerase alpha catalytic subunit; DNA polymerase alpha catalytic subunit DNA polymerase alpha catalytic subunit p180 POLA1 POLA DNA pol alpha; DNA polymerase alpha catalytic subunit p180; DNA polymerase alpha p180 subunit; polymerase (DNA directed), alpha 1, catalytic subunit; polymerase (DNA) alpha 1, catalytic subunit; polymerase (DNA-directed), alpha (70kD)
Gene Aliases: NSX; p180; POLA; POLA1
UniProt ID: (Human) P09884
Entrez Gene ID: (Human) 5422
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.