Invitrogen
Manufacturer Code:PA5113132
Catalog # PIPA5113132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Protein O-glucosyltransferase 1 protein (132-392AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9630046K23Rik; 9630046K23Rik C3orf9 CAP10-like 46 kDa protein CAP10-like protein Clp46 hCLP46 hRumi KDELC family like 1 KDELCL1 KTEL (Lys-Tyr-Glu-Leu) containing 1 KTEL motif-containing protein 1 KTELC1 MDS010 MDSRP Myelodysplastic syndromes relative protein O-glucosyltransferase Rumi homolog POGLUT1 Protein O-glucosyltransferase 1 Protein O-xylosyltransferase Protein O-xylosyltransferase POGLUT1 RGD1306248 Rumi UNQ490/PRO1006 Wing-shaped neural plate protein Wsnp x 010 protein; CAP10-like 46 kDa protein; hCLP46; hRumi; KDELC family like 1; KTEL (Lys-Tyr-Glu-Leu) containing 1; KTEL motif-containing protein 1; Myelodysplastic syndromes relative protein; O-glucosyltransferase Rumi homolog; Protein O-glucosyltransferase 1; protein O-xylosyltransferase; Protein O-xylosyltransferase POGLUT1; x 010 protein
Gene Aliases: C3orf9; CLP46; hCLP46; KDELCL1; KTELC1; MDS010; MDSRP; POGLUT1; Rumi; UNQ490/PRO1006
UniProt ID: (Human) Q8NBL1
Entrez Gene ID: (Human) 56983
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.