Proteintech
Manufacturer Code:177641AP150UL
Catalog # 50-173-1775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | POFUT2 Fusion Protein Ag12148 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C21orf80 FUT13 KIAA0958 O FucT 2 Peptide O fucosyltransferase 2 POFUT2 protein O fucosyltransferase 2; GDP-fucose protein O-fucosyltransferase 2; O-FucT-2; Peptide-O-fucosyltransferase 2
Gene Aliases: 2310011G23Rik; AI256847; BC003494; C21orf80; FUT13; KIAA0958; POFUT2
UniProt ID: (Human) Q9Y2G5, (Mouse) Q8VHI3
Entrez Gene ID: (Human) 23275, (Rat) 309686, (Mouse) 80294
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.