Invitrogen
Manufacturer Code:PA570431
Catalog # PIPA570431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human POFUT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FUT12 O-FUT O-Fuc-T O-FucT-1 GDP-fucose protein O-fucosyltransferase 1 o-fucosyltransferase protein peptide-O-fucosyltransferase 1; GDP-fucose protein O-fucosyltransferase 1; o-fucosyltransferase protein; O-FucT-1; O-FucT-1a; O-FucT-1b; Peptide-O-fucosyltransferase 1; pofut1 {ECO:0000312|RGD:1303001}
Gene Aliases: DDD2; FUT12; KIAA0180; mKIAA0180; O-Fuc-T; O-FucT-1; O-FUT; OFUCT1; POFUT1; zgc:110472
UniProt ID: (Human) Q9H488, (Mouse) Q91ZW2, (Rat) Q6EV70
Entrez Gene ID: (Human) 23509, (Dog) 609881, (Bovine) 360194, (Horse) 100068638, (Rabbit) 108175392, (Guinea pig) 100733160, (Mouse) 140484, (Rat) 311551, (Zebrafish) 403029
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.