Invitrogen
Manufacturer Code:PA562980
Catalog # PIPA562980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human POFUT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FUT12 O-FUT O-Fuc-T O-FucT-1 GDP-fucose protein O-fucosyltransferase 1 o-fucosyltransferase protein peptide-O-fucosyltransferase 1; GDP-fucose protein O-fucosyltransferase 1; o-fucosyltransferase protein; O-FucT-1; Peptide-O-fucosyltransferase 1
Gene Aliases: DDD2; FUT12; KIAA0180; O-Fuc-T; O-FucT-1; O-FUT; OFUCT1; POFUT1
UniProt ID: (Human) Q9H488
Entrez Gene ID: (Human) 23509
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.