Invitrogen
Manufacturer Code:PA5101606
Catalog # PIPA5101606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human POFUT1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDD2 FUT12 GDP-fucose protein O-fucosyltransferase 1 KIAA0180 mKIAA0180 o-fucosyltransferase protein O-Fuc-T OFUCT1 O-FucT-1 O-FUT Peptide-O-fucosyltransferase 1 POFUT1 pofut1 {ECO:0000312 protein O-fucosyltransferase 1 RGD:1303001}; GDP-fucose protein O-fucosyltransferase 1; o-fucosyltransferase protein; O-FucT-1; Peptide-O-fucosyltransferase 1; pofut1 {ECO:0000312|RGD:1303001}
Gene Aliases: DDD2; FUT12; KIAA0180; mKIAA0180; O-Fuc-T; O-FucT-1; O-FUT; OFUCT1; POFUT1
UniProt ID: (Human) Q9H488, (Mouse) Q91ZW2, (Rat) Q6EV70
Entrez Gene ID: (Human) 23509, (Mouse) 140484, (Rat) 311551
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.