Novus Biologicals
Manufacturer Code:NBP222466
Catalog # NBP222466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues R(392) K T I G N T P M E K L L C D M F K N(410) of human PNR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nuclear receptor subfamily 2 group E member 3; Nuclear receptor subfamily 2 group E member 3 nuclear receptor subfamily 2 group E member 3 photoreceptor-specific nuclear receptor PNRESCS rd7 Retina-specific nuclear receptor RNR RP37MGC49976; nuclear receptor subfamily 2, group E, member 3; Photoreceptor-specific nuclear receptor; Retina-specific nuclear receptor
Gene Aliases: ESCS; NR2E3; PNR; rd7; RNR; RP37
UniProt ID: (Human) Q9Y5X4
Entrez Gene ID: (Human) 10002
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.