Novus Biologicals
Manufacturer Code:NBP297714AF700
Catalog # NB046964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PNPT1 fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'-5' RNA exonuclease OLD35; DKFZp762K1914 EC 2.7.7 EC 2.7.7.8 mitochondrial old-35 OLD35 PNPase PNPase 1 PNPase old-35 Polynucleotide phosphorylase 1 Polynucleotide phosphorylase-like protein polyribonucleotide nucleotidyltransferase 1; PNPase 1; PNPase old-35; Polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; Polyribonucleotide nucleotidyltransferase 1, mitochondrial
Gene Aliases: COXPD13; DFNB70; old-35; OLD35; PNPASE; PNPT1
UniProt ID: (Human) Q8TCS8
Entrez Gene ID: (Human) 87178
Molecular Function: RNA binding protein esterase exoribonuclease hydrolase nuclease nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.