Novus Biologicals
Manufacturer Code:NBP197941FR
Catalog # NBP197941FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C7 |
Immunogen | Recombinant human PNPO (57-261aa) purified from E. coli |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 1.4.3.5 FLJ10535 PDXPO pyridoxal 5'-phosphate synthase pyridoxamine 5'-phosphate oxidase Pyridoxamine-phosphate oxidase pyridoxine 5'-phosphate oxidase pyridoxine-5'-phosphate oxidase; epididymis secretory protein Li 302; pyridoxal 5'-phosphate synthase; Pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; Pyridoxine-5'-phosphate oxidase
Gene Aliases: HEL-S-302; PDXPO; PNPO
UniProt ID: (Human) Q9NVS9
Entrez Gene ID: (Human) 55163
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.