Invitrogen
Manufacturer Code:PA544902
Catalog # PIPA544902
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PNPO |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 302; pyridoxal 5'-phosphate synthase; Pyridoxamine 5&prime-phosphate oxidase FLJ10535 PDXPO; Pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; Pyridoxine-5'-phosphate oxidase
Gene Aliases: AI415282; HEL-S-302; PDXPO; PNPO
UniProt ID: (Human) Q9NVS9, (Bovine) Q5E9K3, (Rat) O88794, (Mouse) Q91XF0
Entrez Gene ID: (Horse) 100055489, (Rabbit) 100349278, (Human) 55163, (Bovine) 512573, (Dog) 480540, (Guinea pig) 100718098, (Rat) 64533, (Mouse) 103711
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.