Novus Biologicals
Manufacturer Code:NBP174214
Catalog # NBP174214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to the C terminal of Pnpla6. Immunizing peptide sequence DGCADGEESDCLTEYEEDAGPDCSRDEGGSPEGASPSTASEVEEEKSTLR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1.5 neuropathy target esterase NTENTEMND patatin-like phospholipase domain containing 6 Patatin-like phospholipase domain-containing protein 6 SPG39 sws; Neuropathy target esterase; patatin-like phospholipase domain containing 6; Patatin-like phospholipase domain-containing protein 6
Gene Aliases: BNHS; iPLA2delta; LNMS; NTE; NTEMND; OMCS; PNPLA6; SPG39; sws
UniProt ID: (Human) Q8IY17
Entrez Gene ID: (Human) 10908
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.