Invitrogen
Manufacturer Code:PA5117066
Catalog # PIPA5117066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 377-481 of human PNPLA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase PNPLA3; 1-acylglycerol-3-phosphate O-acyltransferase PNPLA3 acylglycerol O-acyltransferase Acylglycerol transacylase Adiponutrin Adpn C22orf20 Calcium-independent phospholipase A2-epsilon iPLA(2)epsilon iPLA2epsilon iPLA2-epsilon Lysophosphatidic acid acyltransferase patatin like phospholipase domain containing 3 patatin-like phospholipase domain containing 3 patatin-like phospholipase domain-containing protein 3 Pnpla3; acylglycerol O-acyltransferase; Acylglycerol transacylase; Adiponutrin; ADPN; Calcium-independent phospholipase A2-epsilon; iPLA2-epsilon; iPLA2epsilon; Lysophosphatidic acid acyltransferase; patatin-like phospholipase domain containing 3; Patatin-like phospholipase domain-containing protein 3
Gene Aliases: ADPN; C22orf20; iPLA(2)epsilon; PNPLA3
UniProt ID: (Human) Q9NST1, (Mouse) Q91WW7
Entrez Gene ID: (Human) 80339, (Rat) 362972, (Mouse) 116939
Molecular Function:
acyltransferase
hydrolase
lipase
phospholipase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.