Abnova
Manufacturer Code:PAB7591
Catalog # 89-133-396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human PNPLA3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of PNPLA3.
For Research Use Only
Protein Aliases: 1-acylglycerol-3-phosphate O-acyltransferase PNPLA3; acylglycerol O-acyltransferase; Acylglycerol transacylase; Adiponutrin; ADPN; C22orf20; Calcium-independent phospholipase A2-epsilon; iPLA(2)epsilon; iPLA2-epsilon; iPLA2epsilon; Lysophosphatidic acid acyltransferase; patatin-like phospholipase domain containing 3; Patatin-like phospholipase domain-containing protein 3
Gene Aliases: ADPN; C22orf20; iPLA(2)epsilon; PNPLA3
UniProt ID: (Human) Q9NST1
Entrez Gene ID: (Human) 80339
Molecular Function:
acyltransferase
hydrolase
lipase
phospholipase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.