Invitrogen
Manufacturer Code:PA5113223
Catalog # PIPA5113223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Patatin-like phospholipase domain-containing protein 1 protein (101-300AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARCI10 dJ50J22.1 patatin like phospholipase domain containing 1 patatin-like phospholipase domain containing 1 Patatin-like phospholipase domain-containing protein 1 PNPLA1; Omega-hydroxyceramide transacylase; patatin-like phospholipase domain containing 1; Patatin-like phospholipase domain-containing protein 1
Gene Aliases: ARCI10; dJ50J22.1; PNPLA1
UniProt ID: (Human) Q8N8W4, (Mouse) Q3V1D5
Entrez Gene ID: (Human) 285848, (Mouse) 433091
Molecular Function:
acyltransferase
hydrolase
lipase
phospholipase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.