Invitrogen
Manufacturer Code:PA5115775
Catalog # PIPA5115775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PNP corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AL024301 AU015798 epididymis secretory sperm binding protein Li 156an HEL-S-156an inosine phosphorylase inosine phosphorylase {ECO:0000250 Inosine-guanosine phosphorylase NP Np-1 Np-2 nucleoside phosphorylase PNP PNP {ECO:0000250 Pnp1 PRO1837 PUNP Purine nucleoside phosphorylase purine nucleoside phosphorylase {ECO:0000250 purine-nucleoside phosphorylase purine-nucleoside phosphorylase 1 purine-nucleoside:orthophosphate ribosyltransferase UniProtKB:P00491}; epididymis secretory sperm binding protein Li 156an; HEL-S-156an; Inosine phosphorylase; Inosine-guanosine phosphorylase; PNP; Purine nucleoside phosphorylase; purine-nucleoside:orthophosphate ribosyltransferase
Gene Aliases: NP; PNP; PRO1837; PUNP
UniProt ID: (Human) P00491
Entrez Gene ID: (Human) 4860
Molecular Function:
phosphorylase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.