Invitrogen
Manufacturer Code:PA588845
Catalog # PIPA588845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 301-470 of human PNLIPRP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytotoxic T lymphocyte lipase; cytotoxic T-lymphocyte lipase; Galactolipase; Pancreatic lipase-related protein 2; PL-RP2; Secretory glycoprotein GP-3; Triacylglycerol lipase
Gene Aliases: PLRP2; Pnliprp2
UniProt ID: (Rat) P54318, (Mouse) P17892
Entrez Gene ID: (Rat) 117554, (Mouse) 18947
Molecular Function: esterase hydrolase lipase phospholipase storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.