Abnova
Manufacturer Code:H00005408A01
Catalog # 89-105-157
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 333 a.a. &bsim 435 a.a) partial recombinant protein with GST tag.,PNLIPRP2 (NP_005387 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PNLIPRP2.
For Research Use Only
Protein Aliases: Cytotoxic T lymphocyte lipase; Galactolipase; Pancreatic lipase-related protein 2; PL-RP2; PLRP2; Triacylglycerol lipase
Gene Aliases: PLRP2; PNLIPRP2
UniProt ID: (Human) P54317
Entrez Gene ID: (Human) 5408
Molecular Function: esterase hydrolase lipase phospholipase storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.