Proteintech
Manufacturer Code:262181AP150UL
Catalog # 50-173-5261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PNLIPRP2 Fusion Protein Ag23967 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytotoxic T lymphocyte lipase; Galactolipase; Pancreatic lipase-related protein 2; PL RP2 PLRP2 PNLIPRP2; PL-RP2; Triacylglycerol lipase
Gene Aliases: PLRP2; PNLIPRP2
UniProt ID: (Human) P54317
Entrez Gene ID: (Human) 5408
Molecular Function:
esterase
hydrolase
lipase
phospholipase
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.