Invitrogen
Manufacturer Code:PA587217
Catalog # PIPA587217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PNLIP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pancreatic triacylglycerol lipase; Pancreatic triacylglycerol lipase PL PTL Pancreatic lipase PNLIP; pancreatic triglyceride lipase; PL
Gene Aliases: 1810007A24Rik; PANLI; PL; PNLIP; PNLIPD; PTL
UniProt ID: (Human) P16233, (Rat) P27657, (Mouse) Q6P8U6
Entrez Gene ID: (Human) 5406, (Rat) 25702, (Mouse) 69060
Molecular Function: esterase hydrolase lipase phospholipase storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.