Invitrogen
Manufacturer Code:PA569358
Catalog # PIPA569358
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse PNKD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain protein 17; hydrolase PNKD-like protein; MR-1; Myofibrillogenesis regulator 1; paroxysmal nonkinesigenic dyskinesia; paroxysmal nonkinesiogenic dyskinesia; Paroxysmal nonkinesiogenic dyskinesia protein; PNKD, MBL domain containing; Probable hydrolase PNKD; Trans-activated by hepatitis C virus core protein 2
Gene Aliases: 2210013N15Rik; 2810403H05Rik; AI854243; BRP17; DYT8; FKSG19; FPD1; KIAA1184; KIPP1184; MNCb-5687; MR-1; MR1; PDC; PKND1; PNKD; TAHCCP2; UNQ2491/PRO5778
UniProt ID: (Bovine) A7YY46, (Human) Q8N490, (Mouse) Q69ZP3
Entrez Gene ID: (Horse) 106780846, (Bovine) 616561, (Rabbit) 100350646, (Goat) 100861377, (Dog) 610423, (Human) 25953, (Rat) 100188944, (Mouse) 56695, (Guinea pig) 100722586
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.