Invitrogen
Manufacturer Code:PA576992
Catalog # PIPA576992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PNKP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2'(3')-polynucleotidase; Bifunctional polynucleotide phosphatase/kinase; Bifunctional polynucleotide phosphatase/kinase DNA 5&prime-kinase/3&prime-phosphatase Polynucleotide kinase-3&prime-phosphatase PNKP; DNA 5'-kinase/3'-phosphatase; Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP); Polynucleotide 3'-phosphatase; Polynucleotide 5'-hydroxyl-kinase; Polynucleotide kinase-3'-phosphatase
Gene Aliases: 1810009G08Rik; AOA4; EIEE10; MCSZ; PNK; PNKP
UniProt ID: (Human) Q96T60
Entrez Gene ID: (Human) 11284, (Mouse) 59047, (Rat) 308576
Molecular Function:
hydrolase
kinase
nucleotide kinase
nucleotide phosphatase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.