Novus Biologicals
Manufacturer Code:NB1001067
Catalog # NB1001067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence EPRLGRLYCQFSEG corresponding to C-Terminus according to NP_009185.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 2'(3')-polynucleotidase; Bifunctional polynucleotide phosphatase/kinase; DNA 5'-kinase/3'-phosphatase; DNA 5'-kinase/3'-phosphatase EIEE10 Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP) MCSZ PNKbifunctional polynucleotide phosphatase/kinase polynucleotide kinase 3'-phosphatase Polynucleotide kinase-3'-phosphatase; Homo sapiens polynucleotide kinase 3'-phosphatase (PNKP); Polynucleotide 3'-phosphatase; Polynucleotide 5'-hydroxyl-kinase; Polynucleotide kinase-3'-phosphatase
Gene Aliases: AOA4; EIEE10; MCSZ; PNK; PNKP
UniProt ID: (Human) Q96T60
Entrez Gene ID: (Human) 11284
Molecular Function:
hydrolase
kinase
nucleotide kinase
nucleotide phosphatase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.