Invitrogen
Manufacturer Code:PA565589
Catalog # PIPA565589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human PNAS4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DeSI-2; desumoylating isopeptidase 1; Desumoylating isopeptidase 2; desumoylating isopeptidase 2 C1orf121 DESI DESI1 DeSI-2 FAM152A PNAS-4 PPPDE1; Deubiquitinase DESI2; family with sequence similarity 152, member A; PPPDE peptidase domain-containing protein 1; Protein FAM152A
Gene Aliases: C1orf121; CGI-146; DESI; DeSI-2; DESI1; DESI2; FAM152A; PNAS-4; PPPDE1
UniProt ID: (Human) Q9BSY9
Entrez Gene ID: (Human) 51029
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.