Invitrogen
Manufacturer Code:PA542033
Catalog # PIPA542033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human DESI2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DeSI-2; desumoylating isopeptidase 1; Desumoylating isopeptidase 2; desumoylating isopeptidase 2 C1orf121 DESI DESI1 DeSI-2 FAM152A PNAS-4 PPPDE1; Deubiquitinase DESI2; fam152a; family with sequence similarity 152, member A; pnas4; PPPDE peptidase domain containing 1; PPPDE peptidase domain-containing protein 1; Protein FAM152A
Gene Aliases: 5830417C01Rik; C1orf121; cb238; CGI-146; DESI; DeSI-2; DESI1; DESI2; FAM152A; fi12d12; PNAS-4; PPPDE1; sb:cb238; si:ch211-132e22.3; wu:fi12d12; zgc:100860
UniProt ID: (Human) Q9BSY9, (Rat) Q5XIT6, (Mouse) Q9D291, (Zebrafish) Q6DC39
Entrez Gene ID: (Dog) 490377, (Horse) 100062283, (Rabbit) 100351684, (Human) 51029, (Bovine) 519650, (Rat) 289277, (Guinea pig) 100724590, (Mouse) 78825, (Zebrafish) 445138
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.