Invitrogen
Manufacturer Code:PA520763
Catalog # PIPA520763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide near the carboxy terminus of human PNAS4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI-146 C1orf121 FAM152A PNAS-4 PPPDE peptidase domain-containing protein 1 family with sequence similarity 152 member A; DeSI-2; desumoylating isopeptidase 1; Desumoylating isopeptidase 2; Deubiquitinase DESI2; family with sequence similarity 152, member A; PPPDE peptidase domain containing 1; PPPDE peptidase domain-containing protein 1; Protein FAM152A
Gene Aliases: 5830417C01Rik; C1orf121; CGI-146; DESI; DeSI-2; DESI1; DESI2; FAM152A; PNAS-4; PPPDE1
UniProt ID: (Human) Q9BSY9, (Rat) Q5XIT6, (Mouse) Q9D291
Entrez Gene ID: (Human) 51029, (Rat) 289277, (Mouse) 78825
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.