Invitrogen
Manufacturer Code:PA5109629
Catalog # PIPA5109629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human DESI2 (NP_057160.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5830417C01Rik BOS_15659 C1orf121 cb238 CGI-146 DESI desi1 Desi2 DeSI-2 Desumoylating isopeptidase 2 Deubiquitinase DESI2 fam152a fi12d12 PNAS-4 PPPDE peptidase domain-containing protein 1 pppde1 Protein FAM152A sb:cb238 si:ch211-132e22.3 wu:fi12d12 zgc:100860; DeSI-2; desumoylating isopeptidase 1; Desumoylating isopeptidase 2; Deubiquitinase DESI2; family with sequence similarity 152, member A; PPPDE peptidase domain-containing protein 1; Protein FAM152A
Gene Aliases: C1orf121; CGI-146; DESI; DeSI-2; DESI1; DESI2; FAM152A; PNAS-4; PPPDE1
UniProt ID: (Human) Q9BSY9
Entrez Gene ID: (Human) 51029
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.