Novus Biologicals
Manufacturer Code:NBP273484H
Catalog # NBP273484H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F9 |
Immunogen | Full length recombinant protein of human PMVK/phosphomevalonate kinase (NP_006547) produced in HEK293T cells. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: hPMK HUMPMKI phosphomevalonate kinase PMKA PMKase PMKEC 2.7.4.2 PMKI; Phosphomevalonate kinase; PMKase; testis tissue sperm-binding protein Li 95mP
Gene Aliases: HUMPMKI; PMK; PMKA; PMKASE; PMKI; PMVK; POROK1
UniProt ID: (Human) Q15126
Entrez Gene ID: (Human) 10654
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.