OriGene
Manufacturer Code:TA503448
Catalog # 50-167-4342
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D1 |
Immunogen | Full length human recombit protein of human PMVK produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HUMPMKI PMK PMKA PMKASE POROK1; Phosphomevalonate kinase; PMKase; testis tissue sperm-binding protein Li 95mP
Gene Aliases: 1110011E12Rik; 2900002L22Rik; AV001327; HUMPMKI; PMK; PMKA; PMKASE; PMKI; PMVK; POROK1
UniProt ID: (Human) Q15126, (Mouse) Q9D1G2
Entrez Gene ID: (Human) 10654, (Mouse) 68603
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.