Invitrogen
Manufacturer Code:MA529487
Catalog # PIMA529487
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 3 |
Immunogen | Recombit Human PMVK/phosphomevalonate kinase Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Phosphomevalonate kinase; Phosphomevalonate kinase PMKase PMVK PMKI; PMKase; testis tissue sperm-binding protein Li 95mP
Gene Aliases: HUMPMKI; PMK; PMKA; PMKASE; PMKI; PMVK; POROK1
UniProt ID: (Human) Q15126
Entrez Gene ID: (Human) 10654
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.