Novus Biologicals
Manufacturer Code:NBP267069
Catalog # NBP267069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SY08-09 |
Immunogen | Full length protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: DNA mismatch repair protein PMS2; DNA mismatch repair protein PMS2 EC 3.1 H_DJ0042M02.9 HNPCC4postmeiotic segregation increased (S. cerevisiae) 2 mismatch repair endonuclease PMS2 PMS1 protein homolog 2 PMS2 postmeiotic segregation increased 2 (S. cerevisiae) PMSL2PMS2CL; Mismatch repair endonuclease PMS2; PMS1 homolog 2, mismatch repair protein; PMS1 protein homolog 2; PMS2 postmeiotic segregation increased 2; postmeiotic segregation increased 2 nirs variant 6
Gene Aliases: HNPCC4; MLH4; PMS2; PMS2CL; PMSL2
UniProt ID: (Human) P54278
Entrez Gene ID: (Human) 5395
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.